Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
Chromosome Location
Chr.2: 233354507 - 233472098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003648.2 NP_003639.2 14 - 15 1838 77
NM_152879.2 NP_690618.2 15 - 16 1903 77
XM_005246107.3 XP_005246164.1 13 - 14 1570 77
XM_005246108.1 XP_005246165.1 13 - 14 1724 77
XM_005246110.3 XP_005246167.1 4 - 5 1575 77
XM_011512028.2 XP_011510330.1 15 - 16 3362 77
XM_011512029.2 XP_011510331.1 15 - 16 3363 77
XM_011512030.2 XP_011510332.1 15 - 16 1959 77
XM_011512031.2 XP_011510333.1 15 - 16 1833 77
XM_011512032.2 XP_011510334.1 15 - 16 1817 77
XM_011512033.2 XP_011510335.1 15 - 16 3364 77
XM_011512035.1 XP_011510337.1 13 - 14 1704 77
XM_011512037.2 XP_011510339.1 13 - 14 1808 77
XM_011512039.2 XP_011510341.1 15 - 16 3364 77
XM_011512040.2 XP_011510342.1 13 - 14 1740 77
XM_017005138.1 XP_016860627.1 15 - 16 2241 77
XM_017005139.1 XP_016860628.1 13 - 14 1733 77
XM_017005140.1 XP_016860629.1 15 - 16 1835 77
GenBank mRNA AB078966.1 - 15 - 16 1903 77
AK307426.1 - 3 - 4 461 77
AK308846.1 - 13 - 14 1724 77
AK311044.1 - 13 - 14 1657 77
D63479.3 - 15 - 16 1844 77
D73409.1 - 14 - 15 1839 77

View Full Product Details