Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
Chromosome Location
Chr.2: 233354507 - 233472098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003648.2 NP_003639.2 17 - 18 2212 109
NM_152879.2 NP_690618.2 18 - 19 2277 109
XM_005246107.3 XP_005246164.1 16 - 17 1944 109
XM_005246108.1 XP_005246165.1 16 - 17 2098 109
XM_005246110.3 XP_005246167.1 7 - 8 1949 109
XM_011512028.2 XP_011510330.1 18 - 19 3736 109
XM_011512029.2 XP_011510331.1 18 - 19 3737 109
XM_011512030.2 XP_011510332.1 18 - 19 2333 109
XM_011512031.2 XP_011510333.1 18 - 19 2207 109
XM_011512032.2 XP_011510334.1 18 - 19 2191 109
XM_011512033.2 XP_011510335.1 18 - 19 3738 109
XM_011512035.1 XP_011510337.1 16 - 17 2078 109
XM_011512037.2 XP_011510339.1 16 - 17 2182 109
XM_011512039.2 XP_011510341.1 18 - 19 3738 109
XM_011512040.2 XP_011510342.1 16 - 17 2114 109
XM_017005138.1 XP_016860627.1 18 - 19 2615 109
XM_017005139.1 XP_016860628.1 16 - 17 2107 109
XM_017005140.1 XP_016860629.1 18 - 19 2209 109
GenBank mRNA AB078966.1 - 18 - 19 2277 109
AK307426.1 - 6 - 7 835 109
AK308846.1 - 16 - 17 2098 109
AK311044.1 - 16 - 17 2031 109
D63479.3 - 18 - 19 2218 109
D73409.1 - 17 - 18 2213 109

View Full Product Details