The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003648.2 | NP_003639.2 | 28 - 29 | 3509 | 91 |
NM_152879.2 | NP_690618.2 | 29 - 30 | 3574 | 91 | |
XM_005246107.3 | XP_005246164.1 | 27 - 28 | 3241 | 91 | |
XM_005246108.1 | XP_005246165.1 | 27 - 28 | 3395 | 91 | |
XM_005246110.3 | XP_005246167.1 | 18 - 19 | 3246 | 91 | |
XM_011512028.2 | XP_011510330.1 | 29 - 30 | 5033 | 91 | |
XM_011512030.2 | XP_011510332.1 | 29 - 30 | 3630 | 91 | |
XM_011512031.2 | XP_011510333.1 | 29 - 30 | 3504 | 91 | |
XM_011512032.2 | XP_011510334.1 | 29 - 30 | 3488 | 91 | |
XM_011512035.1 | XP_011510337.1 | 27 - 28 | 3375 | 91 | |
XM_011512037.2 | XP_011510339.1 | 27 - 28 | 3479 | 91 | |
XM_011512040.2 | XP_011510342.1 | 27 - 28 | 3411 | 91 | |
XM_017005138.1 | XP_016860627.1 | 29 - 30 | 3912 | 91 | |
XM_017005139.1 | XP_016860628.1 | 27 - 28 | 3404 | 91 | |
XM_017005140.1 | XP_016860629.1 | 29 - 30 | 3506 | 91 | |
GenBank mRNA | AB078966.1 | - | 29 - 30 | 3574 | 91 |
BC006561.2 | - | 3 - 4 | 363 | 91 | |
D63479.3 | - | 29 - 30 | 3515 | 91 | |
D73409.1 | - | 28 - 29 | 3507 | 91 |