Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
Chromosome Location
Chr.2: 233354507 - 233472098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003648.2 NP_003639.2 3 - 4 405 69
NM_152879.2 NP_690618.2 4 - 5 470 69
XM_005246107.3 XP_005246164.1 2 - 3 137 69
XM_005246108.1 XP_005246165.1 2 - 3 291 69
XM_011512028.2 XP_011510330.1 4 - 5 1929 69
XM_011512029.2 XP_011510331.1 4 - 5 1930 69
XM_011512030.2 XP_011510332.1 4 - 5 526 69
XM_011512031.2 XP_011510333.1 4 - 5 400 69
XM_011512032.2 XP_011510334.1 4 - 5 384 69
XM_011512033.2 XP_011510335.1 4 - 5 1931 69
XM_011512035.1 XP_011510337.1 2 - 3 271 69
XM_011512037.2 XP_011510339.1 2 - 3 375 69
XM_011512039.2 XP_011510341.1 4 - 5 1931 69
XM_011512040.2 XP_011510342.1 2 - 3 307 69
XM_017005138.1 XP_016860627.1 4 - 5 808 69
XM_017005139.1 XP_016860628.1 2 - 3 300 69
XM_017005140.1 XP_016860629.1 4 - 5 402 69
GenBank mRNA AB078966.1 - 4 - 5 470 69
AK308846.1 - 2 - 3 291 69
AK311044.1 - 2 - 3 224 69
D63479.3 - 4 - 5 411 69
D73409.1 - 3 - 4 406 69

View Full Product Details