Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAG
Entrez Gene ID
4099
Gene Name
myelin associated glycoprotein
Gene Aliases
GMA, S-MAG, SIGLEC-4A, SIGLEC4A, SPG75
Chromosome Location
Chr.19: 35292086 - 35313807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002361.3 NP_002352.1 3 - 4 243 61
NM_080600.2 NP_542167.1 3 - 4 243 61
GenBank mRNA AK094545.1 - 3 - 4 168 61
AK222649.1 - 3 - 4 168 61
AK222680.1 - 3 - 4 146 61
AK223562.1 - 3 - 4 204 61
BC053347.1 - 3 - 4 135 61
BC093045.1 - 3 - 4 195 61
BI755065.1 - 3 - 4 193 61
DC331168.1 - 3 - 4 243 61
M29273.1 - 3 - 4 165 61

View Full Product Details