The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139285.3 | NP_644814.1 | 1 - 2 | 409 | 58 |
GenBank mRNA | AF508784.1 | - | 1 - 2 | 409 | 58 |
AF508785.1 | - | 1 - 2 | 409 | 58 |