The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 4 - 5 | 479 | 72 |
NM_001136232.2 | NP_001129704.1 | 3 - 4 | 333 | 72 | |
NM_001278946.1 | NP_001265875.1 | 3 - 4 | 301 | 72 | |
NM_030673.3 | NP_109598.2 | 4 - 5 | 335 | 72 | |
NM_183352.2 | NP_899195.1 | 3 - 4 | 301 | 72 | |
XM_005265379.2 | XP_005265436.1 | 4 - 5 | 352 | 72 | |
XM_017007019.1 | XP_016862508.1 | 4 - 5 | 389 | 72 | |
XM_017007020.1 | XP_016862509.1 | 3 - 4 | 196 | 72 | |
XM_017007021.1 | XP_016862510.1 | 3 - 4 | 206 | 72 | |
GenBank mRNA | AB209554.1 | - | 2 - 3 | 148 | 72 |
AF052155.1 | - | 3 - 4 | 333 | 72 | |
AK223019.1 | - | 3 - 4 | 186 | 72 | |
AK294662.1 | - | 3 - 4 | 206 | 72 | |
AK301999.1 | - | 4 - 5 | 403 | 72 | |
BC002634.2 | - | 4 - 5 | 216 | 72 | |
BC091506.1 | - | 3 - 4 | 211 | 72 | |
BE621430.1 | - | 3 - 4 | 287 | 72 | |
CN344033.1 | - | 3 - 4 | 313 | 72 | |
EF036494.1 | - | 3 - 4 | 200 | 72 | |
L09260.1 | - | 3 - 4 | 178 | 72 |