Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB7A
Entrez Gene ID
7879
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
PRO2706, RAB7
Chromosome Location
Chr.3: 128726136 - 128814798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004637.5 NP_004628.4 4 - 5 633 95
GenBank mRNA AF498942.1 - 3 - 4 401 95
AK225177.1 - 3 - 4 387 95
AK225691.1 - 3 - 4 380 95
AK290721.1 - 4 - 5 537 95
BC008721.2 - 4 - 5 576 95
BC013728.1 - 3 - 4 416 95
BX537775.1 - 2 - 3 349 95

View Full Product Details