The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281476.2 | NP_001268405.1 | 10 - 11 | 2342 | 154 |
NM_001322049.1 | NP_001308978.1 | 9 - 10 | 2273 | 154 | |
NR_104020.2 | 10 - 11 | 2342 | 154 | ||
XM_017004912.1 | XP_016860401.1 | 6 - 7 | 2353 | 154 | |
GenBank mRNA | AK055892.1 | - | 10 - 11 | 2253 | 154 |
AK130797.1 | - | 10 - 11 | 2266 | 154 |