The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242837.1 | NP_001229766.1 | 2 - 3 | 348 | 140 |
NM_012305.3 | NP_036437.1 | 2 - 3 | 348 | 140 | |
XM_011519930.2 | XP_011518232.1 | 2 - 3 | 309 | 140 | |
GenBank mRNA | AB020706.2 | - | 2 - 3 | 135 | 140 |
AK001986.1 | - | 2 - 3 | 277 | 140 | |
AK223558.1 | - | 2 - 3 | 276 | 140 | |
AK293763.1 | - | 2 - 3 | 207 | 140 | |
AK299364.1 | - | 2 - 3 | 126 | 140 | |
BC006155.1 | - | 2 - 3 | 276 | 140 | |
BG705558.1 | - | 2 - 3 | 316 | 140 | |
BI462785.1 | - | 2 - 3 | 326 | 140 | |
DB461570.1 | - | 2 - 3 | 348 | 140 |