Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL6B
Entrez Gene ID
51412
Gene Name
actin like 6B
Gene Aliases
ACTL6, BAF53B, arpNalpha
Chromosome Location
Chr.7: 100643097 - 100656461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016188.4 NP_057272.1 2 - 3 211 89
NR_134539.1 2 - 3 211 89
GenBank mRNA AB015906.1 - 2 - 3 175 89
AF041475.1 - 2 - 3 174 89
AK314940.1 - 2 - 3 211 89
BC020944.1 - 2 - 3 198 89

View Full Product Details