Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL6B
Entrez Gene ID
51412
Gene Name
actin like 6B
Gene Aliases
ACTL6, BAF53B, arpNalpha
Chromosome Location
Chr.7: 100643097 - 100656461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016188.4 NP_057272.1 9 - 10 929 62
NR_134539.1 9 - 10 929 62
GenBank mRNA AB015906.1 - 9 - 10 893 62
AF041475.1 - 9 - 10 892 62
AK314940.1 - 9 - 10 929 62
BC020944.1 - 9 - 10 916 62

View Full Product Details