The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006255.4 | NP_006246.2 | 2 - 3 | 1086 | 121 |
XM_011536956.1 | XP_011535258.1 | 2 - 3 | 968 | 121 | |
XM_011536957.1 | XP_011535259.1 | 2 - 3 | 968 | 121 | |
XM_017021459.1 | XP_016876948.1 | 2 - 3 | 968 | 121 | |
GenBank mRNA | AB451313.1 | - | 2 - 3 | 427 | 121 |
AK290183.1 | - | 2 - 3 | 805 | 121 | |
AX775933.1 | - | 2 - 3 | 815 | 121 | |
BC037268.1 | - | 2 - 3 | 815 | 121 | |
M55284.1 | - | 2 - 3 | 590 | 121 |