The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286839.1 | NP_001273768.1 | 15 - 16 | 1937 | 70 |
NM_005766.3 | NP_005757.1 | 15 - 16 | 2116 | 70 | |
XM_011521046.2 | XP_011519348.1 | 15 - 16 | 2116 | 70 | |
XM_017020312.1 | XP_016875801.1 | 15 - 16 | 1860 | 70 | |
XM_017020313.1 | XP_016875802.1 | 14 - 15 | 2853 | 70 | |
GenBank mRNA | AB008430.1 | - | 15 - 16 | 1746 | 70 |
BC041595.1 | - | 15 - 16 | 1963 | 70 | |
BC071592.1 | - | 15 - 16 | 1937 | 70 |