The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005766.3 | NP_005757.1 | 19 - 20 | 2688 | 100 |
XM_017020312.1 | XP_016875801.1 | 19 - 20 | 2432 | 100 | |
GenBank mRNA | AB008430.1 | - | 19 - 20 | 2318 | 100 |
BC041595.1 | - | 19 - 20 | 2535 | 100 |