Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
Chromosome Location
Chr.19: 58346806 - 58353499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130786.3 NP_570602.2 5 - 6 969 76
GenBank mRNA AB073611.1 - 5 - 6 962 76
AK055885.1 - 4 - 5 1557 76
BC035719.1 - 5 - 6 1016 76
EU794585.1 - 5 - 6 961 76

View Full Product Details