Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
Chromosome Location
Chr.6: 32968660 - 32981505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113182.2 NP_001106653.1 6 - 7 2438 288
NM_001199455.1 NP_001186384.1 5 - 6 2128 288
NM_001199456.1 NP_001186385.1 5 - 6 841 288
NM_001291986.1 NP_001278915.1 7 - 8 2530 288
NM_005104.3 NP_005095.1 6 - 7 2525 288
GenBank mRNA AK056504.1 - 6 - 7 2396 288
AL832722.1 - 7 - 8 2465 288
BC000477.2 - 5 - 6 1497 288
BC063840.1 - 5 - 6 993 288
BX647233.1 - 5 - 6 841 288
BX648109.1 - 6 - 7 2439 288
D42040.1 - 6 - 7 2525 288
M80613.1 - 5 - 6 1860 288
X62083.1 - 5 - 6 1860 288

View Full Product Details