The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015384.4 | NP_056199.2 | 25 - 26 | 5508 | 90 |
NM_133433.3 | NP_597677.2 | 25 - 26 | 5508 | 90 | |
XM_005248280.3 | XP_005248337.1 | 25 - 26 | 5497 | 90 | |
XM_005248282.4 | XP_005248339.3 | 19 - 20 | 4999 | 90 | |
XM_006714467.2 | XP_006714530.1 | 25 - 26 | 5497 | 90 | |
XM_006714468.2 | XP_006714531.1 | 24 - 25 | 5299 | 90 | |
XM_011514015.1 | XP_011512317.1 | 25 - 26 | 5497 | 90 | |
XM_017009329.1 | XP_016864818.1 | 25 - 26 | 5497 | 90 | |
XM_017009330.1 | XP_016864819.1 | 17 - 18 | 7771 | 90 | |
XM_017009331.1 | XP_016864820.1 | 24 - 25 | 3871 | 90 | |
GenBank mRNA | AJ627032.1 | - | 25 - 26 | 5477 | 90 |
AJ640137.1 | - | 25 - 26 | 5477 | 90 | |
KJ807791.1 | - | 11 - 12 | 1295 | 90 |