The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015384.4 | NP_056199.2 | 35 - 36 | 6754 | 127 |
NM_133433.3 | NP_597677.2 | 35 - 36 | 6754 | 127 | |
XM_005248280.3 | XP_005248337.1 | 35 - 36 | 6743 | 127 | |
XM_005248282.4 | XP_005248339.3 | 29 - 30 | 6245 | 127 | |
XM_006714467.2 | XP_006714530.1 | 35 - 36 | 6743 | 127 | |
XM_006714468.2 | XP_006714531.1 | 34 - 35 | 6545 | 127 | |
XM_011514015.1 | XP_011512317.1 | 35 - 36 | 6743 | 127 | |
XM_017009329.1 | XP_016864818.1 | 35 - 36 | 6743 | 127 | |
XM_017009330.1 | XP_016864819.1 | 27 - 28 | 9017 | 127 | |
XM_017009331.1 | XP_016864820.1 | 34 - 35 | 5117 | 127 | |
GenBank mRNA | AJ627032.1 | - | 35 - 36 | 6723 | 127 |
AJ640137.1 | - | 35 - 36 | 6723 | 127 | |
AK023416.1 | - | 3 - 4 | 402 | 127 | |
KJ807791.1 | - | 19 - 20 | 2295 | 127 |