The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319164.1 | NP_001306093.1 | 6 - 7 | 706 | 124 |
NM_080574.3 | NP_542141.1 | 6 - 7 | 788 | 124 | |
GenBank mRNA | AF432917.1 | - | 5 - 6 | 640 | 124 |
AK291854.1 | - | 6 - 7 | 788 | 124 | |
AY359055.1 | - | 6 - 7 | 706 | 124 | |
BC065726.1 | - | 6 - 7 | 715 | 124 |