The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128595.2 | NP_001122067.1 | 4 - 5 | 682 | 128 |
NM_001128596.2 | NP_001122068.1 | 4 - 5 | 801 | 128 | |
NM_001289134.1 | NP_001276063.1 | 4 - 5 | 682 | 128 | |
NM_152332.5 | NP_689545.1 | 4 - 5 | 870 | 128 | |
GenBank mRNA | AK093876.1 | - | 4 - 5 | 778 | 128 |
AK126359.1 | - | 4 - 5 | 639 | 128 | |
BC040503.1 | - | 4 - 5 | 683 | 128 | |
BX647828.1 | - | 4 - 5 | 604 | 128 |