The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000924.3 | NP_000915.1 | 3 - 4 | 622 | 87 |
NM_001165975.2 | NP_001159447.1 | 2 - 3 | 310 | 87 | |
NM_001288769.1 | NP_001275698.1 | 2 - 3 | 242 | 87 | |
XM_011538456.1 | XP_011536758.1 | 4 - 5 | 572 | 87 | |
XM_017019432.1 | XP_016874921.1 | 2 - 3 | 141 | 87 | |
XM_017019433.1 | XP_016874922.1 | 2 - 3 | 829 | 87 | |
GenBank mRNA | AB209200.1 | - | 3 - 4 | 391 | 87 |
AJ401609.1 | - | 2 - 3 | 169 | 87 | |
AK296422.1 | - | 2 - 3 | 242 | 87 | |
AK302931.1 | - | 2 - 3 | 297 | 87 | |
BC032226.1 | - | 3 - 4 | 395 | 87 | |
BP342896.1 | - | 2 - 3 | 289 | 87 | |
DC298452.1 | - | 2 - 3 | 439 | 87 | |
DC392024.1 | - | 2 - 3 | 297 | 87 | |
EU668337.1 | - | 3 - 4 | 391 | 87 | |
U56976.1 | - | 2 - 3 | 263 | 87 | |
U86078.1 | - | 3 - 4 | 342 | 87 |