Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFXN2
Entrez Gene ID
118980
Gene Name
sideroflexin 2
Gene Aliases
-
Chromosome Location
Chr.10: 102714518 - 102739189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178858.4 NP_849189.1 9 - 10 934 58
XM_011539261.2 XP_011537563.1 10 - 11 935 58
XM_011539262.2 XP_011537564.1 10 - 11 1055 58
XM_011539263.2 XP_011537565.1 11 - 12 1099 58
XM_011539264.2 XP_011537566.1 10 - 11 961 58
GenBank mRNA AF462052.1 - 9 - 10 889 58
AK055711.1 - 7 - 8 696 58
AK308246.1 - 9 - 10 838 58
BC022091.2 - 9 - 10 934 58

View Full Product Details