The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178819.3 | NP_848934.1 | 8 - 9 | 1844 | 72 |
XM_011544392.2 | XP_011542694.1 | 9 - 10 | 1967 | 72 | |
XM_011544393.1 | XP_011542695.1 | 8 - 9 | 1856 | 72 | |
XM_011544394.1 | XP_011542696.1 | 8 - 9 | 1893 | 72 | |
XM_017013042.1 | XP_016868531.1 | 9 - 10 | 1994 | 72 | |
XM_017013043.1 | XP_016868532.1 | 9 - 10 | 1865 | 72 | |
GenBank mRNA | AF406612.1 | - | 8 - 9 | 1832 | 72 |
AL834255.1 | - | 4 - 5 | 329 | 72 | |
AY358670.1 | - | 7 - 8 | 981 | 72 | |
AY513610.1 | - | 7 - 8 | 1064 | 72 | |
BC051377.1 | - | 7 - 8 | 1024 | 72 | |
BC061884.1 | - | 7 - 8 | 1024 | 72 |