Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr.18: 23689443 - 23955066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000227.4 NP_000218.3 29 - 30 3905 109
NM_001127717.2 NP_001121189.2 65 - 66 8780 109
NM_001127718.2 NP_001121190.2 28 - 29 3737 109
NM_198129.2 NP_937762.2 66 - 67 8948 109
XM_011525978.2 XP_011524280.1 66 - 67 8975 109
XM_011525979.2 XP_011524281.1 66 - 67 8966 109
XM_011525980.2 XP_011524282.1 66 - 67 8957 109
XM_011525981.2 XP_011524283.1 65 - 66 8843 109
XM_011525982.2 XP_011524284.1 64 - 65 8678 109
XM_017025743.1 XP_016881232.1 49 - 50 6659 109
XM_017025744.1 XP_016881233.1 39 - 40 5340 109
GenBank mRNA AB107369.1 - 66 - 67 8707 109
AB208853.1 - 21 - 22 2933 109
AY327114.1 - 28 - 29 3712 109
AY327115.1 - 66 - 67 8707 109
AY327116.1 - 65 - 66 8539 109
BC043618.1 - 23 - 24 3285 109
L34155.1 - 29 - 30 3847 109
X84900.1 - 31 - 32 4126 109

View Full Product Details