Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLOD3
Entrez Gene ID
8985
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Gene Aliases
LH3
Chromosome Location
Chr.7: 101205977 - 101217730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084.4 NP_001075.1 11 - 12 1688 64
GenBank mRNA AF046889.1 - 11 - 12 1448 64
AF068229.1 - 11 - 12 1293 64
AK297559.1 - 10 - 11 1432 64
AK298537.1 - 12 - 13 1376 64
AK312743.1 - 11 - 12 1446 64
AL049952.1 - 1 - 2 113 64
AY220458.1 - 11 - 12 1296 64
BC011674.2 - 11 - 12 1545 64
BC018946.2 - 6 - 7 610 64

View Full Product Details