Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLOD3
Entrez Gene ID
8985
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Gene Aliases
LH3
Chromosome Location
Chr.7: 101205977 - 101217730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084.4 NP_001075.1 14 - 15 2065 60
GenBank mRNA AF046889.1 - 14 - 15 1825 60
AF068229.1 - 14 - 15 1670 60
AK075338.1 - 14 - 15 1913 60
AK312743.1 - 14 - 15 1823 60
AY220458.1 - 14 - 15 1673 60
BC011674.2 - 14 - 15 1922 60
BC018946.2 - 9 - 10 987 60

View Full Product Details