The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084.4 | NP_001075.1 | 5 - 6 | 1076 | 96 |
GenBank mRNA | AF046889.1 | - | 5 - 6 | 836 | 96 |
AF068229.1 | - | 5 - 6 | 681 | 96 | |
AK297559.1 | - | 5 - 6 | 657 | 96 | |
AK298537.1 | - | 6 - 7 | 763 | 96 | |
AK312743.1 | - | 5 - 6 | 834 | 96 | |
AY220458.1 | - | 5 - 6 | 684 | 96 | |
BC011674.2 | - | 5 - 6 | 933 | 96 |