The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178078.1 | NP_001171549.1 | 13 - 14 | 1840 | 82 |
NM_001178079.1 | NP_001171550.1 | 13 - 14 | 1766 | 82 | |
NM_001178080.1 | NP_001171551.1 | 12 - 13 | 1684 | 82 | |
NM_001178081.1 | NP_001171552.1 | 11 - 12 | 1545 | 82 | |
NM_003153.4 | NP_003144.3 | 13 - 14 | 1821 | 82 | |
NR_033659.1 | 12 - 13 | 1682 | 82 | ||
XM_006719575.2 | XP_006719638.1 | 14 - 15 | 1818 | 82 | |
GenBank mRNA | AB103089.1 | - | 7 - 8 | 993 | 82 |
AK290431.1 | - | 13 - 14 | 1769 | 82 | |
AK291044.1 | - | 13 - 14 | 1767 | 82 | |
AK299094.1 | - | 12 - 13 | 1618 | 82 | |
AK308751.1 | - | 13 - 14 | 1661 | 82 | |
AK312471.1 | - | 13 - 14 | 1687 | 82 | |
AK316142.1 | - | 12 - 13 | 1628 | 82 | |
AK316236.1 | - | 12 - 13 | 1459 | 82 | |
BC075852.1 | - | 13 - 14 | 1622 | 82 | |
BT007380.1 | - | 12 - 13 | 1515 | 82 | |
U16031.1 | - | 13 - 14 | 1680 | 82 |