The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178078.1 | NP_001171549.1 | 15 - 16 | 2073 | 72 |
NM_001178079.1 | NP_001171550.1 | 15 - 16 | 1999 | 72 | |
NM_001178080.1 | NP_001171551.1 | 14 - 15 | 1917 | 72 | |
NM_001178081.1 | NP_001171552.1 | 13 - 14 | 1778 | 72 | |
NM_003153.4 | NP_003144.3 | 15 - 16 | 2054 | 72 | |
NR_033659.1 | 14 - 15 | 1915 | 72 | ||
XM_006719575.2 | XP_006719638.1 | 16 - 17 | 2051 | 72 | |
XM_011538703.2 | XP_011537005.1 | 16 - 17 | 2128 | 72 | |
XM_011538704.2 | XP_011537006.1 | 17 - 18 | 2139 | 72 | |
XM_011538705.2 | XP_011537007.1 | 17 - 18 | 2249 | 72 | |
XM_011538706.2 | XP_011537008.1 | 17 - 18 | 2105 | 72 | |
XM_011538707.2 | XP_011537009.1 | 16 - 17 | 2090 | 72 | |
XM_011538708.2 | XP_011537010.1 | 15 - 16 | 1932 | 72 | |
XM_011538709.2 | XP_011537011.1 | 16 - 17 | 1966 | 72 | |
GenBank mRNA | AB103089.1 | - | 9 - 10 | 1226 | 72 |
AK290431.1 | - | 15 - 16 | 2002 | 72 | |
AK291044.1 | - | 15 - 16 | 2000 | 72 | |
AK298721.1 | - | 13 - 14 | 1598 | 72 | |
AK299094.1 | - | 14 - 15 | 1851 | 72 | |
AK308751.1 | - | 15 - 16 | 1894 | 72 | |
AK312471.1 | - | 15 - 16 | 1920 | 72 | |
AK316142.1 | - | 14 - 15 | 1861 | 72 | |
AK316236.1 | - | 14 - 15 | 1692 | 72 | |
AY615283.1 | - | 3 - 4 | 72 | ||
BC075852.1 | - | 15 - 16 | 1855 | 72 | |
BT007380.1 | - | 14 - 15 | 1748 | 72 | |
U16031.1 | - | 15 - 16 | 1913 | 72 |