The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000435.2 | NP_000426.2 | 14 - 15 | 2370 | 105 |
XM_005259924.4 | XP_005259981.1 | 14 - 15 | 2309 | 105 | |
GenBank mRNA | AB209447.1 | - | 14 - 15 | 2303 | 105 |
U97669.1 | - | 14 - 15 | 2372 | 105 |