Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 11 - 12 1351 64
XM_011529935.1 XP_011528237.1 11 - 12 1360 64
XM_011529936.1 XP_011528238.1 11 - 12 1360 64
XM_011529937.1 XP_011528239.1 11 - 12 1360 64
XM_011529938.1 XP_011528240.1 10 - 11 1243 64
XM_011529939.1 XP_011528241.1 11 - 12 1360 64
XM_011529940.1 XP_011528242.1 11 - 12 1360 64
XM_011529941.1 XP_011528243.1 11 - 12 1360 64
XM_011529942.2 XP_011528244.1 11 - 12 1551 64
XM_011529943.1 XP_011528245.1 11 - 12 1360 64
XM_011529944.1 XP_011528246.1 11 - 12 1360 64
XM_011529945.2 XP_011528247.1 11 - 12 1362 64
XM_011529948.2 XP_011528250.1 7 - 8 732 64
XM_011529949.1 XP_011528251.1 6 - 7 669 64
XM_011529950.1 XP_011528252.1 11 - 12 1360 64
XM_011529952.1 XP_011528254.1 6 - 7 806 64
XM_011529954.1 XP_011528256.1 1 - 2 84 64
XM_011529955.1 XP_011528257.1 11 - 12 1360 64
XM_011529956.2 XP_011528258.1 11 - 12 1360 64
XM_017028633.1 XP_016884122.1 11 - 12 1360 64
GenBank mRNA BC025400.1 - 11 - 12 1367 64
BC070111.1 - 6 - 7 808 64

View Full Product Details