The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152513.3 | NP_689726.3 | 9 - 10 | 1121 | 119 |
XM_011529935.1 | XP_011528237.1 | 9 - 10 | 1130 | 119 | |
XM_011529936.1 | XP_011528238.1 | 9 - 10 | 1130 | 119 | |
XM_011529937.1 | XP_011528239.1 | 9 - 10 | 1130 | 119 | |
XM_011529939.1 | XP_011528241.1 | 9 - 10 | 1130 | 119 | |
XM_011529940.1 | XP_011528242.1 | 9 - 10 | 1130 | 119 | |
XM_011529941.1 | XP_011528243.1 | 9 - 10 | 1130 | 119 | |
XM_011529942.2 | XP_011528244.1 | 9 - 10 | 1321 | 119 | |
XM_011529943.1 | XP_011528245.1 | 9 - 10 | 1130 | 119 | |
XM_011529944.1 | XP_011528246.1 | 9 - 10 | 1130 | 119 | |
XM_011529945.2 | XP_011528247.1 | 9 - 10 | 1132 | 119 | |
XM_011529946.1 | XP_011528248.1 | 9 - 10 | 1130 | 119 | |
XM_011529947.1 | XP_011528249.1 | 9 - 10 | 1130 | 119 | |
XM_011529948.2 | XP_011528250.1 | 5 - 6 | 502 | 119 | |
XM_011529949.1 | XP_011528251.1 | 4 - 5 | 439 | 119 | |
XM_011529950.1 | XP_011528252.1 | 9 - 10 | 1130 | 119 | |
XM_011529955.1 | XP_011528257.1 | 9 - 10 | 1130 | 119 | |
XM_011529956.2 | XP_011528258.1 | 9 - 10 | 1130 | 119 | |
XM_017028633.1 | XP_016884122.1 | 9 - 10 | 1130 | 119 | |
GenBank mRNA | BC025400.1 | - | 9 - 10 | 1137 | 119 |