The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_038230.1 | 2 - 3 | 246 | 110 | |
GenBank mRNA | AK096761.1 | - | 2 - 3 | 246 | 110 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | distal-less homeobox 3 | DLX3 | AI4, TDO | -55.1kb | Sense | Chr. - 49990005 - 49995224 on Build GRCh38 |
Downstream | integrin subunit alpha 3 | ITGA3 | CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a | 2.3kb | Anti-sense | Chr. - 50055968 - 50090485 on Build GRCh38 |