Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS23
Entrez Gene ID
6228
Gene Name
ribosomal protein S23
Gene Aliases
S23
Chromosome Location
Chr.5: 82273320 - 82278416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025.4 NP_001016.1 1 - 2 97 100
GenBank mRNA BU594771.1 - 1 - 2 97 100
CF129812.1 - 1 - 2 186 100

View Full Product Details