Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SH2D6
- Entrez Gene ID
-
284948
- Gene Name
- SH2 domain containing 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 85418931 - 85437029 on Build GRCh38
View Full Product Details