Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TICRR
Entrez Gene ID
90381
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C15orf42, SLD3, Treslin
Chromosome Location
Chr.15: 89575587 - 89628023 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308025.1 NP_001294954.1 6 - 7 1673 108
NM_152259.3 NP_689472.3 6 - 7 1676 108
GenBank mRNA DR731358.1 - 1 - 2 108
GQ227787.1 - 6 - 7 1673 108

View Full Product Details