The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052846.1 | NP_443078.1 | 3 - 4 | 736 | 159 |
GenBank mRNA | AB089149.1 | - | 3 - 4 | 736 | 159 |
BC101043.1 | - | 3 - 4 | 512 | 159 | |
BC101044.2 | - | 3 - 4 | 512 | 159 | |
BC101045.1 | - | 3 - 4 | 512 | 159 |