The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320479.1 | NP_001307408.1 | 9 - 10 | 806 | 53 |
XM_017027654.1 | XP_016883143.1 | 10 - 11 | 1114 | 53 | |
XM_017027655.1 | XP_016883144.1 | 10 - 11 | 905 | 53 | |
XM_017027657.1 | XP_016883146.1 | 9 - 10 | 1003 | 53 | |
XM_017027658.1 | XP_016883147.1 | 10 - 11 | 1054 | 53 | |
XM_017027659.1 | XP_016883148.1 | 10 - 11 | 1060 | 53 | |
XM_017027660.1 | XP_016883149.1 | 10 - 11 | 942 | 53 | |
XM_017027661.1 | XP_016883150.1 | 9 - 10 | 1051 | 53 | |
XM_017027662.1 | XP_016883151.1 | 10 - 11 | 912 | 53 | |
GenBank mRNA | AK000652.1 | - | 2 - 3 | 122 | 53 |
AK097430.1 | - | 9 - 10 | 808 | 53 |