Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN2
Entrez Gene ID
645104
Gene Name
clarin 2
Gene Aliases
-
Chromosome Location
Chr.4: 17515165 - 17527104 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079827.2 NP_001073296.1 1 - 2 354 97
GenBank mRNA BC127863.1 - 1 - 2 354 97

View Full Product Details