Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
Chromosome Location
Chr.14: 97925610 - 97978124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK091668.1 - 2 - 3 409 105

View Full Product Details