The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102386.2 | NP_001095856.1 | 5 - 6 | 670 | 92 |
XM_011516124.2 | XP_011514426.1 | 5 - 6 | 1164 | 92 | |
GenBank mRNA | BC147016.1 | - | 5 - 6 | 670 | 92 |
BC147017.1 | - | 5 - 6 | 670 | 92 |