Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
Chromosome Location
Chr.9: 34621360 - 34633112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017363.1 NP_001017363.1 3 - 4 570 59
XM_017014283.1 XP_016869772.1 5 - 6 3656 59

View Full Product Details