Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD3
Entrez Gene ID
84443
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.X: 107522450 - 107605264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032428.1 NP_115804.1 9 - 10 865 68
XM_006724709.2 XP_006724772.1 3 - 4 340 68
XM_011531056.2 XP_011529358.1 7 - 8 841 68
XM_011531057.2 XP_011529359.1 7 - 8 767 68
XM_011531058.2 XP_011529360.1 6 - 7 679 68
XM_017029901.1 XP_016885390.1 7 - 8 1086 68
XM_017029902.1 XP_016885391.1 7 - 8 1086 68

View Full Product Details