Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTS2B
Entrez Gene ID
257313
Gene Name
urotensin 2B
Gene Aliases
U2B, URP, UTS2D
Chromosome Location
Chr.3: 191267155 - 191330536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198152.3 NP_937795.2 6 - 7 990 105
XM_011512631.1 XP_011510933.1 5 - 6 655 105
XM_017006091.1 XP_016861580.1 5 - 6 655 105
GenBank mRNA AB116021.1 - 2 - 3 252 105
AK090630.1 - 6 - 7 989 105
AY321313.1 - 2 - 3 265 105
BC112166.1 - 2 - 3 248 105

View Full Product Details