The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004326.4 | NP_001004326.4 | 4 - 5 | 799 | 89 |
NR_033396.1 | 5 - 6 | 968 | 89 | ||
XM_011545043.2 | XP_011543345.1 | 4 - 5 | 799 | 89 | |
XM_017017774.1 | XP_016873263.1 | 4 - 5 | 799 | 89 | |
XM_017017775.1 | XP_016873264.1 | 2 - 3 | 775 | 89 | |
XM_017017776.1 | XP_016873265.1 | 2 - 3 | 775 | 89 | |
GenBank mRNA | AK131327.1 | - | 4 - 5 | 801 | 89 |
BC146850.1 | - | 4 - 5 | 791 | 89 | |
BM826695.1 | - | 1 - 2 | 131 | 89 | |
DN831899.1 | - | 1 - 2 | 63 | 89 | |
DY654969.1 | - | 2 - 3 | 238 | 89 |