Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
Chromosome Location
Chr.X: 75053172 - 75156340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271696.1 NP_001258625.1 7 - 8 1010 99
NM_001271697.1 NP_001258626.1 6 - 7 890 99
NM_001271698.1 NP_001258627.1 6 - 7 932 99
NM_001271699.1 NP_001258628.1 6 - 7 893 99
NM_004299.4 NP_004290.2 7 - 8 1013 99
GenBank mRNA AF038950.1 - 7 - 8 950 99
AF078777.1 - 7 - 8 930 99
AF133659.1 - 7 - 8 942 99
AK001418.1 - 7 - 8 970 99
AK294657.1 - 6 - 7 874 99
AK307414.1 - 6 - 7 813 99
BC006323.2 - 7 - 8 946 99
BT009918.1 - 7 - 8 945 99
BX537833.1 - 6 - 7 830 99
KU177872.1 - 7 - 8 945 99
KU177873.1 - 7 - 8 945 99

View Full Product Details