Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 19 - 20 3897 87
NM_015052.4 NP_055867.3 21 - 22 4234 87
XM_005249665.3 XP_005249722.1 20 - 21 4016 87
XM_006715670.3 XP_006715733.1 19 - 20 3914 87
XM_006715671.3 XP_006715734.1 19 - 20 3911 87
XM_006715673.3 XP_006715736.1 18 - 19 3809 87
XM_011515220.2 XP_011513522.1 20 - 21 3807 87
XM_011515222.2 XP_011513524.1 19 - 20 3948 87
XM_011515223.2 XP_011513525.1 19 - 20 3749 87
XM_011515224.2 XP_011513526.1 19 - 20 4229 87
XM_011515225.2 XP_011513527.2 17 - 18 3266 87
XM_011515226.2 XP_011513528.1 16 - 17 3490 87
XM_017011882.1 XP_016867371.1 19 - 20 4256 87
XM_017011883.1 XP_016867372.1 20 - 21 4193 87
XM_017011884.1 XP_016867373.1 19 - 20 4253 87
XM_017011885.1 XP_016867374.1 21 - 22 4189 87
XM_017011886.1 XP_016867375.1 20 - 21 4018 87
XM_017011887.1 XP_016867376.1 18 - 19 4154 87
XM_017011888.1 XP_016867377.1 18 - 19 4151 87
XM_017011889.1 XP_016867378.1 19 - 20 4208 87
XM_017011890.1 XP_016867379.1 20 - 21 4018 87
GenBank mRNA AB002320.2 - 21 - 22 4234 87
AB048365.1 - 18 - 19 3586 87
AK294918.1 - 19 - 20 3851 87
AL536075.3 - 5 - 6 529 87
BC151227.1 - 21 - 22 4234 87

View Full Product Details