Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 26 - 27 4781 66
NM_015052.4 NP_055867.3 28 - 29 5118 66
XM_005249665.3 XP_005249722.1 27 - 28 4900 66
XM_006715670.3 XP_006715733.1 26 - 27 4798 66
XM_006715671.3 XP_006715734.1 26 - 27 4795 66
XM_006715673.3 XP_006715736.1 25 - 26 4693 66
XM_011515220.2 XP_011513522.1 27 - 28 4691 66
XM_011515222.2 XP_011513524.1 26 - 27 4832 66
XM_011515223.2 XP_011513525.1 26 - 27 4633 66
XM_011515224.2 XP_011513526.1 26 - 27 5113 66
XM_011515225.2 XP_011513527.2 24 - 25 4150 66
XM_011515226.2 XP_011513528.1 23 - 24 4374 66
XM_017011882.1 XP_016867371.1 26 - 27 5140 66
XM_017011883.1 XP_016867372.1 27 - 28 5077 66
XM_017011884.1 XP_016867373.1 26 - 27 5137 66
XM_017011885.1 XP_016867374.1 28 - 29 5073 66
XM_017011886.1 XP_016867375.1 27 - 28 4902 66
XM_017011887.1 XP_016867376.1 25 - 26 5038 66
XM_017011888.1 XP_016867377.1 25 - 26 5035 66
XM_017011889.1 XP_016867378.1 26 - 27 5092 66
GenBank mRNA AB002320.2 - 28 - 29 5118 66
AB048365.1 - 25 - 26 4470 66
AK294918.1 - 26 - 27 4735 66
BC151227.1 - 28 - 29 5118 66

View Full Product Details