Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 8 - 9 1317 63
NM_015052.4 NP_055867.3 9 - 10 1552 63
XM_005249665.3 XP_005249722.1 8 - 9 1331 63
XM_006715670.3 XP_006715733.1 7 - 8 1229 63
XM_006715671.3 XP_006715734.1 7 - 8 1229 63
XM_006715673.3 XP_006715736.1 7 - 8 1229 63
XM_011515220.2 XP_011513522.1 8 - 9 1122 63
XM_011515222.2 XP_011513524.1 7 - 8 1263 63
XM_011515223.2 XP_011513525.1 7 - 8 1064 63
XM_011515224.2 XP_011513526.1 7 - 8 1544 63
XM_011515225.2 XP_011513527.2 5 - 6 581 63
XM_011515226.2 XP_011513528.1 4 - 5 805 63
XM_017011882.1 XP_016867371.1 7 - 8 1571 63
XM_017011883.1 XP_016867372.1 8 - 9 1508 63
XM_017011884.1 XP_016867373.1 7 - 8 1571 63
XM_017011885.1 XP_016867374.1 9 - 10 1504 63
XM_017011886.1 XP_016867375.1 8 - 9 1336 63
XM_017011887.1 XP_016867376.1 7 - 8 1571 63
XM_017011888.1 XP_016867377.1 7 - 8 1571 63
XM_017011889.1 XP_016867378.1 8 - 9 1628 63
XM_017011890.1 XP_016867379.1 8 - 9 1336 63
GenBank mRNA AB002320.2 - 9 - 10 1552 63
AB048365.1 - 6 - 7 904 63
AK294918.1 - 8 - 9 1271 63
BC151227.1 - 9 - 10 1552 63

View Full Product Details